missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride hydrate, 97%, ca 10% water, Thermo Scientific™

Product Code. 11480014
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11480014 10 g 10g
11490014 50 g 50g
11400024 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11480014

Brand: Thermo Scientific Alfa Aesar A13542.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-53-1
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012800
InChI Key YPFMNHZRNXPYBG-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride
PubChem CID 102623
IUPAC Name ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O

Specifications

Melting Point ∼170°C (decomposition)
Quantity 10 g
Beilstein 3918980
Formula Weight 297.78 (Anhydrous)
Percent Purity 97%
Assay ≈10% water
Chemical Name or Material 1-Benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.