missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methyl-4-piperidone, 99%

Product Code. 10755621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10755621 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10755621 Supplier Thermo Scientific Acros Supplier No. 134040250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 34737-89-8 | C13H17NO | 203.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 34737-89-8
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Molecular Formula C13H17NO
Refractive Index 1.5315 to 1.5335
MDL Number MFCD00044806
Quantity 25 g
Beilstein 21,V,6,483
Synonym 1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
InChI Key OVQAJYCAXPHYNV-UHFFFAOYSA-N
SMILES CC1CN(CCC1=O)CC2=CC=CC=C2
IUPAC Name 1-benzyl-3-methylpiperidin-4-one
Molecular Weight (g/mol) 203.28
PubChem CID 560965
Formula Weight 203.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-methyl-4-piperidone, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Benzyl-3-methyl-4-piperidone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.