missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methylimidazolium chloride, 97%

Product Code. 15436468
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 15436468

Brand: Thermo Scientific Alfa Aesar H59997.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36443-80-8 | C11H13ClN2 | 208.689 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36443-80-8
Molecular Formula C11H13ClN2
Molecular Weight (g/mol) 208.689
MDL Number MFCD07784451
InChI Key FCRZSGZCOGHOGF-UHFFFAOYSA-M
Synonym 1-benzyl-3-methylimidazolium chloride, 1-benzyl-3-methyl-1h-imidazol-3-ium chloride, 1h-imidazolium,1-methyl-3-phenylmethyl-, chloride 1:1, 3-benzyl-1-methylimidazol-1-ium chloride, dsstox_cid_27905, dsstox_rid_82658, dsstox_gsid_47929, ksc576i7l, 1-methyl-3-benzylimidazolium chloride, 1-benzyl-3-methylimidaz-olium chloride
PubChem CID 10560335
IUPAC Name 1-benzyl-3-methylimidazol-3-ium;chloride
SMILES C[N+]1=CN(C=C1)CC2=CC=CC=C2.[Cl-]

Specifications

Quantity 50 g
Sensitivity Moisture sensitive
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 1-Benzyl-3-methylimidazolium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.