missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-piperidone hydrate hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 10637504 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637504 1 g Glass bottle 1g
10515881 5 g Glass bottle 5g
10387661 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10637504 Supplier Thermo Scientific Acros Supplier No. 166920010
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50606-58-1
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD00150096
InChI Key OVWSFXNSJDMRPV-UHFFFAOYSA-N
Synonym 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
PubChem CID 3084924
IUPAC Name 1-benzylpiperidin-3-one;hydrochloride
SMILES Cl.O=C1CCCN(CC2=CC=CC=C2)C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 170°C to 176°C
Color Brown to Cream
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on free base) (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 225.72
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Benzyl-3-piperidone hydrate hydrochloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Benzyl-3-piperidone hydrate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.