missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-piperidone hydrochloride hydrate

Product Code. 11465253
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465253 25 g 25g
11445253 1 g 1g
11455253 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465253 Supplier Thermo Scientific Alfa Aesar Supplier No. A12849.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 50606-58-1 | C12H16ClNO | 225.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50606-58-1
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD00150096
InChI Key OVWSFXNSJDMRPV-UHFFFAOYSA-N
Synonym 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
PubChem CID 3084924
IUPAC Name 1-benzylpiperidin-3-one;hydrochloride
SMILES Cl.O=C1CCCN(CC2=CC=CC=C2)C1

Specifications

Melting Point 105°C to 107°C
Quantity 25 g
Beilstein 3705146
Formula Weight 225.72 (Anhydrous)
Chemical Name or Material 1-Benzyl-3-piperidone hydrochloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.