missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-Boc-piperazine, 99%, Thermo Scientific Chemicals

Product Code. 11373626
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373626 5 g 5g
11363626 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373626 Supplier Thermo Scientific Alfa Aesar Supplier No. L14644.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57260-70-5 | Molecular Formula: C16H24N2O2 | Molecular Weight: 276.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57260-70-5
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD00075603
InChI Key GVHSMUYEAWMYLM-UHFFFAOYSA-N
Synonym 1-benzyl-4-boc-piperazine, 1-boc-4-benzyl piperazine, 1-boc-4-benzyl-piperazine, tert-butyl-4-benzyl-1-piperazine carboxylate, 1-piperazinecarboxylic acid, 4-phenylmethyl-, 1,1-dimethylethyl ester, 4-benzyl-piperazine-1-carboxylic acid tert-butyl ester, maybridge4_002104, pubchem8596, 1-boc-4-benzylpiperazine, acmc-1bz2p
PubChem CID 584330
IUPAC Name tert-butyl 4-benzylpiperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC2=CC=CC=C2)CC1

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
Beilstein 797033
Formula Weight 276.38
Percent Purity 99%
Chemical Name or Material 1-Benzyl-4-Boc-piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.