missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-4-piperidinol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1-Benzyl-4-piperidinol |
| CAS | 4727-72-4 |
| Molecular Formula | C12H17NO |
| Quantity | 10 g |
| Synonym | 1-(Phenylmethyl)-4-piperidinol, 4-Hydroxy-1-benzylpiperidine, 4-Hydroxy-N-benzylpiperidine, N-Benzyl-4-hydroxypiperidine, NSC 72991, 1-(Phenylmethyl)-4-piperidin-ol |
| InChI Formula | InChI=1S/C12H17NO/c14-12-6-8-13(9-7-12)10-11-4-2-1-3-5-11/h1-5,12,14H,6-10H2 |
| SMILES | OC1CCN(Cc2ccccc2)CC1 |
| IUPAC Name | 1-benzylpiperidin-4-ol |
| Molecular Weight (g/mol) | 191.27 |
| Formula Weight | 191.131 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction