missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid, ≥90%, Thermo Scientific™

Product Code. 10170632
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10170632 1 g 1g
10408894 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10170632 Supplier Thermo Scientific Maybridge Supplier No. CC13401DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 117483-89-3
Molecular Formula C17H15NO4
Molecular Weight (g/mol) 297.31
MDL Number MFCD02682032
InChI Key BSOYWTITVKXHLM-UHFFFAOYNA-N
Synonym 1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
PubChem CID 2776400
IUPAC Name 1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid
SMILES OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%+
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 297.31
Percent Purity 90%
Chemical Name or Material 1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.