missing translation for 'onlineSavingsMsg'
Learn More

1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10563002
Change view
Click to view available options
Quantity:
1 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10563002 1 g Amber Glass Bottle 1g
10011474 100 g Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10563002 Supplier Thermo Scientific Maybridge Supplier No. CC07501DA
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10314-98-4
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD01568759
InChI Key URTPNQRAHXRPMP-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl piperidine-4-carboxylic acid, n-cbz-4-piperidinecarboxylic acid, 1-cbz-4-piperidinecarboxylic acid, n-cbz-piperidine-4-carboxylic acid, 1-benzyloxycarbonyl piperidine-4-carboxylic acid, 1-cbz-piperidine-4-carboxylic acid, 1-benzyloxy carbonyl-4-piperidinecarboxylic acid, n-cbz-isonipecotic acid, piperidine-1,4-dicarboxylic acid monobenzyl ester
PubChem CID 736489
IUPAC Name 1-phenylmethoxycarbonylpiperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.29
Percent Purity 97%
Chemical Name or Material 1-[(Benzyloxy)carbonyl]piperidine-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.