missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonyl-3-piperidone, 97+%

Product Code. 15491689
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491689 25 g 25g
15471689 1 g 1g
15481689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491689 Supplier Thermo Scientific Alfa Aesar Supplier No. H64471.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61995-20-8 | C13H15NO3 | 233.27 g/mol

1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61995-20-8
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.27
MDL Number MFCD03001723
InChI Key ALXLNFWWLXCXSK-UHFFFAOYSA-N
Synonym 1-n-cbz-3-piperidone, 1-cbz-3-piperidinone, 1-cbz-3-piperidone, n-cbz-3-piperidone, n-benzyloxycarbonyl-3-piperidone, 1-piperidinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxo-piperidine-1-carboxylic acid benzyl ester, 1-benzyloxycarbonyl-3-piperidone, 3-oxopiperidine-1-carboxylic acid benzyl ester, pubchem24305
PubChem CID 1514169
IUPAC Name benzyl 3-oxopiperidine-1-carboxylate
SMILES O=C(OCC1=CC=CC=C1)N1CCCC(=O)C1

Specifications

Quantity 25 g
Beilstein 1464624
Solubility Information Slightly soluble in water.
Formula Weight 233.27
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1-Benzyloxycarbonyl-3-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.