missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonyl-4-piperidone, 98+%

Product Code. 15463247
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15463247 25 g 25g
11440777 1 g 1g
11450777 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15463247

Brand: Thermo Scientific Alfa Aesar B23181.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19099-93-5 | C13H15NO3 | 233.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19099-93-5
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD00673144
InChI Key VZOVOHRDLOYBJX-UHFFFAOYSA-N
Synonym 1-cbz-4-piperidone, n-benzyloxycarbonyl-4-piperidone, 1-cbz-piperidin-4-one, n-cbz-4-piperidone, benzyl 4-oxo-1-piperidinecarboxylate, 1-benzyloxycarbonyl-4-piperidinone, 1-z-4-piperidone, 1-piperidinecarboxylic acid, 4-oxo-, phenylmethyl ester, 1-carbobenzoxy-4-piperidone, 1-benzyloxycarbonyl-4-piperidone
PubChem CID 643496
IUPAC Name benzyl 4-oxopiperidine-1-carboxylate
SMILES C1CN(CCC1=O)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 38°C to 41°C
Density 1.172
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 233.27
Percent Purity ≥98%
Chemical Name or Material 1-Benzyloxycarbonyl-4-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.