missing translation for 'onlineSavingsMsg'
Learn More

1-(Benzyloxycarbonyl)piperazine, 98%

Product Code. 15452428
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452428 100 mL 100mL
15432428 5 mL 5mL
15442428 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452428 Supplier Thermo Scientific Alfa Aesar Supplier No. H56008.AE
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31166-44-6 | C12H17N2O2 | 221.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31166-44-6
Molecular Formula C12H17N2O2
Molecular Weight (g/mol) 221.28
MDL Number MFCD00274317
InChI Key CTOUWUYDDUSBQE-UHFFFAOYSA-O
Synonym 1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine
PubChem CID 643495
IUPAC Name benzyl piperazine-1-carboxylate
SMILES O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1

Specifications

Density 1.142
Boiling Point 158°C to 161°C (1.4 mmHg)
Refractive Index 1.546
Quantity 100 mL
Beilstein 179500
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 1-(Benzyloxycarbonyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.