missing translation for 'onlineSavingsMsg'
Learn More

1-beta-D-Arabinofuranosyluracil, 99%

Product Code. 15466229
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466229 5 g 5g
15456229 1 g 1g
15476229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466229 Supplier Thermo Scientific Alfa Aesar Supplier No. J64147.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3083-77-0 | C9H12N2O6 | 244.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3083-77-0
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.20
MDL Number MFCD00065998
InChI Key DRTQHJPVMGBUCF-CCXZUQQUSA-N
Synonym 1-2r,3s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione
PubChem CID 57416896
SMILES OC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 5 g
Beilstein 28749
IUPAC Name 1-[(2R,3S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Formula Weight 244.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1-beta-D-Arabinofuranosyluracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.