missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-3,3-difluoropyrrolidine, 98%

Product Code. 15433687
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433687 250 mg 250mg
11367249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433687 Supplier Alfa Aesar™ Supplier No. H27422.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 195447-25-7
Molecular Formula C9H15F2NO2
Molecular Weight (g/mol) 207.221
MDL Number MFCD06797010
InChI Key ULFWNKDACVUZOD-UHFFFAOYSA-N
Synonym 1-boc-3,3-difluoropyrrolidine, acmc-20ahbm, n-t-boc-3,3-difluoropyrrolidine, tert-butyl 3,3-difluoropyrroline-1-carboxylate, 3,3-difluoropyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylicacid, 3,3-difluoro-, 1,1-dimethylethyl ester, 1-pyrrolidinecarboxylic acid, 3,3-difluoro-, 1,1-dimethylethyl ester
PubChem CID 18542424
IUPAC Name tert-butyl 3,3-difluoropyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)(F)F

Specifications

Refractive Index 1.42
Quantity 250 mg
Formula Weight 207.22
Percent Purity 98%
Chemical Name or Material 1-Boc-3,3-difluoropyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.