missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-aminoazetidine, 95%

Product Code. p-4812589 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15299636

Brand: Thermo Scientific Acros 451180010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 193269-78-2 | C8H16N2O2 | 172.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193269-78-2
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
InChI Key WPGLRFGDZJSQGI-UHFFFAOYSA-N
Synonym 1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
PubChem CID 1516507
IUPAC Name tert-butyl 3-aminoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)N

Specifications

Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material 1-BOC-3-aminoazetidine
compliance-icons
Product Identifier
  • 1-Boc-3-aminoazetidine, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.