missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-aminoazetidine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-Boc-3-aminoazetidine |
| CAS | 193269-78-2 |
| Molecular Formula | C8H16N2O2 |
| Quantity | 250 mg |
| Synonym | 3-Amino-1-azetidinecarboxylic Acid 1,1-Dimethylethyl Ester, tert-Butyl 3-Amino-1-azetidinecarboxylate, 1,1-Dimethylethyl 3-Aminoazetidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-aminoazetidine |
| InChI Formula | InChI=1S/C8H16N2O2/c1-8(2,3)12-7(11)10-4-6(9)5-10/h6H,4-5,9H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)N1CC(N)C1 |
| IUPAC Name | tert-butyl 3-aminoazetidine-1-carboxylate |
| Molecular Weight (g/mol) | 172.22 |
| Formula Weight | 172.1212 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction