missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-aminopyrrolidine, 96%

Product Code. p-7030506
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15463847

Brand: Thermo Scientific Alfa Aesar H29231.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 186550-13-0 | C9H19N2O2 | 187.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186550-13-0
Molecular Formula C9H19N2O2
Molecular Weight (g/mol) 187.26
MDL Number MFCD01861220
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-O
Synonym 1-boc-3-aminopyrrolidine, 3-amino-1-boc-pyrrolidine, +/--3-amino-1-n-boc-pyrrolidine, 3-amino-1-n-boc-pyrrolidine, 3-amino-1-tert-butoxycarbonyl pyrrolidine, +/--1-boc-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, n-boc-3-aminopyrrolidine, 1-bocap
PubChem CID 2756370
IUPAC Name tert-butyl 3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC[C@@H]([NH3+])C1

Specifications

Density 1.022
Refractive Index 1.471
Quantity 250 mg
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 186.25
Percent Purity 96%
Chemical Name or Material (+/-)-1-Boc-3-aminopyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.