missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-(bromomethyl)azetidine, 95%

Product Code. 15219766 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15219766 1 g 1g
15229766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15219766 Supplier Thermo Scientific Acros Supplier No. 452160010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 253176-93-1 | C9H16BrNO2 | 250.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253176-93-1
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD16556174
InChI Key PUKCUGDJEPVLPR-UHFFFAOYSA-N
Synonym 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate
PubChem CID 53350331
IUPAC Name tert-butyl 3-(bromomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CBr)C1

Specifications

Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 250.13
Percent Purity 95%
Chemical Name or Material 1-BOC-3-(bromomethyl)azetidine
compliance-icons
Product Identifier
  • 1-Boc-3-(bromomethyl)azetidine, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.