missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(ethylamino)azetidine, 95%, Thermo Scientific Chemicals

Product Code. p-7038878
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15404338

Brand: Thermo Scientific Alfa Aesar H57941.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454703-23-2
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD11520619
InChI Key ZXVCULTZOWFSRS-UHFFFAOYSA-N
Synonym tert-butyl 3-ethylamino azetidine-1-carboxylate, 1-boc-3-ethylamino azetidine, 1-boc-3-ethylaminoazetidine, 1-n-boc-3-ethylamino azetidine, 1-azetidinecarboxylic acid,3-ethylamino-,1,1-dim, 1-azetidinecarboxylic acid,3-ethylamino-,1,1-dimethylethyl ester, 1-azetidinecarboxylic acid, 3-ethylamino-, 1,1-dimethylethyl ester 9ci
PubChem CID 45089534
IUPAC Name tert-butyl 3-(ethylamino)azetidine-1-carboxylate
SMILES CCNC1CN(C1)C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 200.28
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material 1-Boc-3-(ethylamino)azetidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.