missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%

Product Code. 15429527
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429527 5 g 5g
15439527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429527 Supplier Thermo Scientific Alfa Aesar Supplier No. H52784.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 116574-71-1 | C11H21NO3 | 215.293 g/mol

(±)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

Solubility
Soluble in methanol and dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116574-71-1
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
MDL Number MFCD03094733
InChI Key OJCLHERKFHHUTB-UHFFFAOYSA-N
Synonym n-boc-piperidine-3-methanol, tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, 1-boc-3-piperidinemethanol, n-boc-3-piperidinemethanol, 3-hydroxymethyl-1-n-boc-piperidine, n-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethyl piperidine, tert-butyl 3-hydroxymethyl tetrahydro-1 2h-pyridinecarboxylate, +/--n-boc-3-piperidine methanol
PubChem CID 2763851
IUPAC Name tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

Melting Point 77°C to 81°C
Quantity 5 g
Solubility Information Soluble in methanol and dimethylformamide(DMF).
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material (+/-)-1-Boc-3-(hydroxymethyl)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.