missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxypiperidine, 97%

Product Code. 11348546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348546 5 g 5g
11338546 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348546 Supplier Thermo Scientific Alfa Aesar Supplier No. L17600.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85275-45-2 | C10H19NO3 | 201.266 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85275-45-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD02093938
InChI Key UIJXHKXIOCDSEB-UHFFFAOYSA-N
Synonym 1-boc-3-hydroxypiperidine, 1-n-boc-3-hydroxy-piperidine, 1-boc-3-hydroxy-piperidine, n-boc-3-piperidinol, n-boc-3-hydroxypiperidine, 1-tert-butoxycarbonyl-3-hydroxy piperidine, boc-3-hydroxypiperidine, n-tert-butoxycarbonyl-3-piperidinol, n-boc-3-hydroxy piperidine, 3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 545699
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Beilstein 4180859
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material 1-Boc-3-hydroxypiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.