missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-methylazetidine, 95%, Thermo Scientific Chemicals

Product Code. 15493958
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493958 500 mg 500mg
15483958 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493958 Supplier Thermo Scientific Alfa Aesar Supplier No. H57496.ME
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314984-00-3 | C10H15NO2 | 181.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314984-00-3
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD17016080
InChI Key UENGYBYGCXKNRF-UHFFFAOYSA-N
Synonym 1-boc-3-ethynylazetidine, 3-ethynyl-1-azetidinecarboxylic acid tert-butyl ester, 1-boc-3-ethynyl-azetidine, tert-butyl3-ethynylazetidine-1-carboxylate, 1-azetidinecarboxylic acid, 3-ethynyl-, 1,1-dimethylethyl ester
PubChem CID 54349237
IUPAC Name tert-butyl 3-ethynylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C#C

Specifications

Quantity 500 mg
Beilstein 15408998
Formula Weight 171.24
Percent Purity 95%
Chemical Name or Material 1-Boc-3-methylazetidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.