missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-chloro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15400089
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400089 1 g 1g
15468296 250 mg 250mg
15410089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400089 Supplier Thermo Scientific Alfa Aesar Supplier No. H63407.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 193902-80-6
Molecular Formula C15H20ClN3O4
Molecular Weight (g/mol) 341.792
InChI Key HTMRIXVUKSIEPV-UHFFFAOYSA-N
Synonym tert-butyl 4-2-chloro-4-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-chloro-4-nitrophenyl piperazine, 3-chloro-4-4-t-butoxycarbonyl-1-piperazinyl nitrobenzene, 4-2-chloro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-chloro-4-nitrophenyl piperazine-1-carboxylic acid t-butyl ester
PubChem CID 18783805
IUPAC Name tert-butyl 4-(2-chloro-4-nitrophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Especificaciones

Quantity 1 g
Beilstein 10018753
Formula Weight 341.79
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(2-chloro-4-nitrophenyl)piperazine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.