missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-4-chloropiperidine, 97%

Product Code. 10774882 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10774882 1 g 1g
10048049 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10774882 Supplier Thermo Scientific Acros Supplier No. 435010010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154874-94-9 | C10H18ClNO2 | 219.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154874-94-9
Molecular Formula C10H18ClNO2
Molecular Weight (g/mol) 219.71
MDL Number MFCD04115040
InChI Key NZZWXABIGMMKQL-UHFFFAOYSA-N
Synonym n-boc-4-chloro-piperidine, 1-boc-4-chloropiperidine, 1-boc-4-chloro-piperidine, 4-chloro-piperidine-1-carboxylic acid tert-butyl ester, n-boc-4-chloropiperidine, 1-piperidinecarboxylic acid, 4-chloro-, 1,1-dimethylethyl ester, chloropiperidine, 1-n-boc-4-chloropiperidine, ksc528i4h, 1-n-boc-4-chloro-piperidine
PubChem CID 11138624
IUPAC Name tert-butyl 4-chloropiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Cl)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 219.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-BOC-4-chloropiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 1-BOC-4-chloro-piperidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.