missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-4-Piperidinecarboxaldehyde, 95%, Thermo Scientific™

Product Code. 11956411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11956411 500 mg Glass Bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11956411 Supplier Thermo Scientific Acros Supplier No. 441755000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 137076-22-3
Melting Point 31.0°C to 34.0°C
Boiling Point 120.0°C (2.0 mmHg)
Flash Point 110°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Molecular Formula C11H19NO3
MDL Number MFCD02179019
Quantity 500 mg
Synonym 1-boc-4-piperidinecarboxaldehyde, 1-boc-4-formylpiperidine, 1-tert-butoxycarbonyl-4-piperidinecarboxaldehyde, 1-boc-piperidine-4-carboxaldehyde, n-boc-4-formylpiperidine, n-boc-4-piperidinecarboxaldehyde, 1-tert-butoxycarbonyl-4-formylpiperidine, 4-formyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-piperidine-4-carbaldehyde, 4-formylpiperidine-1-carboxylic acid tert-butyl ester
InChI Key JYUQEWCJWDGCRX-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C=O
IUPAC Name tert-butyl 4-formylpiperidine-1-carboxylate
Molecular Weight (g/mol) 213.28
PubChem CID 1514430
Formula Weight 213.28
Percent Purity 95%
Chemical Name or Material 1-BOC-4-Piperidinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 1-BOC-4-Piperidinecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.