missing translation for 'onlineSavingsMsg'
Learn More

1-(Boc-amino)cyclopropanecarboxylic Acid, TRC

Product Code. 30336899
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30336899 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30336899

Brand: Toronto Research Chemicals TRCB6196451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Boc-amino)cyclopropanecarboxylic Acid
CAS 88950-64-5
Molecular Formula C34 H30 F N3 O2
Quantity 1 g
Synonym 1-[[(1,1-Dimethylethoxy)carbonyl]amino]cyclopropanecarboxylic Acid, 1-(Tert-Butoxycarbonylamino)cyclopropanecarboxylic Acid, 1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic Acid, 1-[N-(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic Acid, N-tert-Butoxycarbonyl 1-amino-1-cyclopropanecarboxylic Acid
InChI Formula InChI=1S/C34H30FN3O2/c1-40-32(39)33(25-35,37-23-27-14-6-2-7-15-27)22-31-24-38(26-36-31)34(28-16-8-3-9-17-28,29-18-10-4-11-19-29)30-20-12-5-13-21-30/h2-21,23-24,26H,22,25H2,1H3/b37-23+
SMILES COC(=O)C(CF)(Cc1cn(cn1)C(c2ccccc2)(c3ccccc3)c4ccccc4)\N=C\c5ccccc5
IUPAC Name methyl 2-[(E)-benzylideneamino]-2-(fluoromethyl)-3-(1-tritylimidazol-4-yl)propanoate
Molecular Weight (g/mol) 531.619
Formula Weight 531.232
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.