missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-homopiperazine, 98%

Product Code. p-7047145
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378656 1 g 1g
11388656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378656 Supplier Thermo Scientific Alfa Aesar Supplier No. L17728.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112275-50-0 | C10H20N2O2 | 200.282 g/mol

1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-homopiperazine is used in synthesis of potent anticoagulant. It is also used to produce 4-(7,8,9,10-tetrahydro-6H-cyclohepta[b]quinolin-11-yl)-[1,4]diazepane-1-carboxylic acid tert-butyl ester by reaction with 11-bromo-7,8,9,10-tetrahydro-6H-cyclohepta[b]quinoline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112275-50-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD00276987
InChI Key WDPWEXWMQDRXAL-UHFFFAOYSA-N
Synonym 1-boc-homopiperazine, 1-boc-hexahydro-1,4-diazepine, 1,4 diazepane-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl homopiperazine, 1-boc-1,4-diazepane, n-boc-homopiperazine, boc-hopiz, boc-homopiperazine, n-tert-butoxycarbonyl homopiperazine, tert-butyl 1-homopiperazinecarboxylate
PubChem CID 2756058
IUPAC Name tert-butyl 1,4-diazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCNCC1

Specifications

Boiling Point 90°C (0.5mmHg)
Refractive Index 1.471
UN Number UN2735
Beilstein 7581789
Solubility Information Not miscible or difficult to mix in water.
Density 1.016
Quantity 1 g
Flash Point >110°C (230°F)
Formula Weight 200.28
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material 1-Boc-homopiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.