missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-imidazole, 98%

Product Code. 11360074
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11360074

Brand: Thermo Scientific Alfa Aesar L00506.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49761-82-2 | C8H12N2O2 | 168.196 g/mol

1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N?-carbonyldiimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.

Solubility
Soluble in hexane, acetone, and DMF.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49761-82-2
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00014497
InChI Key MTBKGWHHOBJMHJ-UHFFFAOYSA-N
Synonym n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p
PubChem CID 521262
IUPAC Name tert-butyl imidazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CN=C1

Specifications

Melting Point 44°C to 48°C
Quantity 25 g
Beilstein 607792
Solubility Information Soluble in hexane,acetone,and DMF.
Formula Weight 168.2
Percent Purity 98%
Chemical Name or Material 1-Boc-imidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.