missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-L-nipecotic acid, 97%

Product Code. 15499327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499327 5 g 5g
15409337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499327 Supplier Thermo Scientific Alfa Aesar Supplier No. H52575.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 88495-54-9 | C11H19NO4 | 229.276 g/mol

1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88495-54-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD02179172
InChI Key NXILIHONWRXHFA-QMMMGPOBSA-N
Synonym s-1-boc-piperidine-3-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, s-n-boc-piperidine-3-carboxylic acid, l-1-boc-nipecotic acid, s-boc-nipecotic acid, boc-s-nipecotic acid, boc-s-nip-oh, s-boc-nip, s-n-boc-nipecotic acid, boc-nip-oh
PubChem CID 6951168
IUPAC Name (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O

Specifications

Melting Point 165°C to 169°C
Quantity 5 g
Optical Rotation +57° (c=1 in Chloroform)
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material 1-Boc-L-nipecotic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.