missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-piperidine, 98%, Thermo Scientific™

Product Code. 11348446
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11348446

Brand: Thermo Scientific Alfa Aesar L17476.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75844-69-8
Molecular Formula C10H19NO2
Molecular Weight (g/mol) 185.267
MDL Number MFCD02093935
InChI Key RQCNHUCCQJMSRG-UHFFFAOYSA-N
Synonym 1-boc-piperidine, n-boc-piperidine, 1-tert-butoxycarbonyl piperidine, 1-piperidinecarboxylic acid, 1,1-dimethylethyl ester, piperidine-1-carboxylic acid tert-butyl ester, tert-butyl piperidinecarboxylate, boc-piperidine, n-boc piperidine, pubchem7258, n-boc tetrahydropyridine
PubChem CID 7010304
IUPAC Name tert-butyl piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1

Specifications

Density 0.965
Boiling Point 130°C to 140°C (15 mmHg)
Flash Point 86°C (187°F)
Refractive Index 1.4535
Quantity 5 g
Beilstein 3604898
Formula Weight 185.27
Percent Purity 98%
Chemical Name or Material 1-Boc-piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.