missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrole-2-boronic acid pinacol ester, 97%

Product Code. 15401259
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401259 1 g 1g
15421259 250 mg 250mg
15411259 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15401259 Supplier Thermo Scientific Alfa Aesar Supplier No. H64171.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1072944-98-9 | C15H24BNO4 | 293.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072944-98-9
Molecular Formula C15H24BNO4
Molecular Weight (g/mol) 293.17
MDL Number MFCD11504960
InChI Key DZQURIUJAGTAFW-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate
PubChem CID 46739108
IUPAC Name tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C

Spécification

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 293.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Boc-pyrrole-2-boronic acid pinacol ester

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.