missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-pyrrole-2-carboxaldehyde, 98%, Thermo Scientific™

Product Code. 15475287
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475287 25 g 25g
15465287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475287 Supplier Thermo Scientific Alfa Aesar Supplier No. H32553.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161282-57-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD03086220
InChI Key NSHLHMBABZKJHP-UHFFFAOYSA-N
Synonym tert-butyl 2-formyl-1h-pyrrole-1-carboxylate, n-boc-pyrrole-2-carboxaldehyde, 2-formyl-pyrrole-1-carboxylic acid tert-butyl ester, acmc-20ak5v, n-boc-pyrrole-2-carbaldehyde, 1-boc-pyrrole-2-carboxaldehyde, 1-t-butoxycarbonyl pyrrole-2-carbaldehyde, tert-butyl 2-methanoylpyrrole-1-carboxylate
PubChem CID 5057005
IUPAC Name tert-butyl 2-formylpyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=CC=C1C=O

Specifications

Melting Point 49°C to 52°C
Flash Point 107.0°C (225°F)
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 1-Boc-pyrrole-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.