missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane, tech. 90%, stab. with sodium carbonate

Product Code. 11337435
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11337435

Brand: Thermo Scientific Alfa Aesar L05406.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54149-17-6 | C5H11BrO2 | 183.05 g/mol

1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store at 4°C. Protect from heat. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54149-17-6
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000238
InChI Key HUXJXNSHCKHFIL-UHFFFAOYSA-N
Synonym 1-bromo-2-2-methoxyethoxy ethane, 1-2-bromoethoxy-2-methoxyethane, 1-bromo-3,6-dioxaheptane, ethane, 1-2-bromoethoxy-2-methoxy, 2-2-methoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-methoxy-ethane, 1-bromo-2-2-methoxy-ethoxy ethane, mpeg2-br, acmc-1alws, 7-bromo-2,5-dioxaheptane
PubChem CID 123532
IUPAC Name 1-(2-bromoethoxy)-2-methoxyethane
SMILES COCCOCCBr

Spécification

Density 1.347
Boiling Point 156°C
Flash Point 73°C (163°F)
Refractive Index 1.455
Quantity 5 g
Beilstein 2069579
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 183.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane, stabilized with sodium carbonate
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis