missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3-dimethyl-benzene, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dimethyl Bromobenzene |
| CAS | 576-23-8 |
| Molecular Formula | C8 H9 Br |
| Quantity | 50 g |
| Synonym | 2,3-Dimethyl Bromobenzene |
| InChI Formula | InChI=1S/C8H9Br/c1-6-4-3-5-8(9)7(6)2/h3-5H,1-2H3 |
| SMILES | CC1=C(C(=CC=C1)Br)C |
| IUPAC Name | 1-bromo-2,3-dimethylbenzene |
| Molecular Weight (g/mol) | 185.0611 |
| Formula Weight | 183.9888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction