missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-butyne, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-bromobut-2-yne |
| CAS | 3355-28-0 |
| Molecular Formula | C4 H5 Br |
| Quantity | 5 g |
| Synonym | 1-Bromo-2-butyne, 1-Bromo-3-methyl-2-butyne, 2-Butyn-1-yl Bromide, 2-Butynyl Bromide, 4-Bromobut-2-yne |
| InChI Formula | InChI=1S/C4H5Br/c1-2-3-4-5/h4H2,1H3 |
| SMILES | CC#CCBr |
| IUPAC Name | 1-bromobut-2-yne |
| Molecular Weight (g/mol) | 132.987 |
| Formula Weight | 131.957 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction