missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane, TRC

Product Code. 30331268
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331268 Supplier Toronto Research Chemicals Supplier No. TRCB68215525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromo-2-chloroethane
CAS 107-04-0
Molecular Formula C2 H4 Br Cl
Quantity 25 g
Synonym 1-Bromo-2-chloroethane, 1-Chloro-2-bromoethane, 2-Bromo-1-chloroethane, 2-Bromoethyl chloride, 2-Chloroethyl bromide, Ethylene chlorobromide, NSC 60186, β-Chloroethyl bromide
InChI Formula InChI=1S/C2H4BrCl/c3-1-2-4/h1-2H2
SMILES ClCCBr
IUPAC Name 1-bromo-2-chloroethane
Molecular Weight (g/mol) 143.41
Formula Weight 141.9185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.