missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbutane, 97%, Thermo Scientific Chemicals

Product Code. 10542851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10542851 25 mL Glass bottle 25mL
10348880 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10542851 Supplier Thermo Scientific Acros Supplier No. 305180250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3814-34-4 | C6H13Br | 165.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3814-34-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.08
MDL Number MFCD00000219
InChI Key KKGUMGWNFARLSL-UHFFFAOYSA-N
Synonym 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane
PubChem CID 77432
IUPAC Name 3-(bromomethyl)pentane
SMILES CCC(CC)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 143°C to 144°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2CHCH2Br
Refractive Index 1.4510 to 1.453
Quantity 25 mL
Specific Gravity 1.17
Formula Weight 165.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-ethylbutane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.