missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluorobenzene, TRC

Product Code. 30335649
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335649 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335649 Supplier Toronto Research Chemicals Supplier No. TRCB68862525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromo-2-fluorobenzene
CAS 1072-85-1
Molecular Formula C6 H4 Br F
Quantity 25 g
Synonym 1-Bromo-2-fluorobenzene, 1-Fluoro-2-bromobenzene, 2-Bromo-1-fluorobenzene, 2-Bromofluorobenzene, 2-Bromophenyl fluoride, 2-Fluoro-1-bromobenzene, 2-Fluorobromobenzene, 2-Fluorophenyl bromide, NSC 59696, o-Bromofluorobenzene, o-Fluorobromobenzene
InChI Formula InChI=1S/C6H4BrF/c7-5-3-1-2-4-6(5)8/h1-4H
SMILES Fc1ccccc1Br
IUPAC Name 1-bromo-2-fluorobenzene
Molecular Weight (g/mol) 175
Formula Weight 173.948
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.