missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylnaphthalene, 90%, technical

Product Code. 10195683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10195683 5 g Glass bottle 5g
10764271 25 g Glass bottle 25g
10143300 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10195683 Supplier Thermo Scientific Acros Supplier No. 154260050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2586-62-1 | C11H9Br | 221.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2586-62-1
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00003871
InChI Key CMIMBQIBIZZZHQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl
PubChem CID 75754
IUPAC Name 1-bromo-2-methylnaphthalene
SMILES CC1=CC=C2C=CC=CC2=C1Br

Specifications

CAS Min % 90.0
CAS Max % 100.0
Color Yellow
Density 1.4100g/mL
Boiling Point 296.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3C10H6Br
Refractive Index 1.6480 to 1.6500
Quantity 5 g
Beilstein 05, 568
Specific Gravity 1.41
Formula Weight 221.1
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylnaphthalene, 90-95%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2-methylnaphthalene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.