missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%

Product Code. 11496696
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496696 250 g 250g
11486696 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496696 Supplier Thermo Scientific Alfa Aesar Supplier No. B23854.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-77-3 | C4H9Br | 137.02 g/mol

1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

Solubility
Slightly soluble in water. Miscible with alcohol, ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-77-3
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000217
InChI Key HLVFKOKELQSXIQ-UHFFFAOYSA-N
Synonym isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
PubChem CID 6555
IUPAC Name 1-bromo-2-methylpropane
SMILES CC(C)CBr

Specifications

Melting Point -118°C
Density 1.255
Boiling Point 90°C to 92°C
Flash Point 18°C (64°F)
Refractive Index 1.435
Quantity 250 g
UN Number UN2342
Beilstein 1730915
Merck Index 14,5135
Solubility Information Slightly soluble in water; Miscible with alcohol,ether.
Formula Weight 137.02
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-methylpropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromo-2-methylpropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.