missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,3-dimethylbutan-2-one, Tech., Thermo Scientific™

Product Code. 10041794
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10041794 10 g 10g
10051794 25 g 25g
10742587 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10041794 Supplier Thermo Scientific Maybridge Supplier No. CD08380EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr

Specifications

CAS Min % 80.0
CAS Max % 100.0
Density 1.3300g/mL
Packaging Amber glass bottle
Linear Formula (CH3)3CCOCH2Br
Quantity 10 g
Formula Weight 179.06
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-Bromo-3, 3-dimethylbutan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.