missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenoxypropane, 98%

Product Code. 11343186
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343186 100 g 100g
11333186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343186 Supplier Thermo Scientific Alfa Aesar Supplier No. L12494.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 588-63-6 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-63-6
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00000256
InChI Key NIDWUZTTXGJFNN-UHFFFAOYSA-N
Synonym 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide
PubChem CID 68522
IUPAC Name 3-bromopropoxybenzene
SMILES BrCCCOC1=CC=CC=C1

Specifications

Melting Point 10°C to 11°C
Density 1.365
Boiling Point 130°C to 134°C (14 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 508978
Formula Weight 215.1
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenoxypropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.