missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluorobutane, 97%

Product Code. 15441469
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441469 25 g 25g
15421469 1 g 1g
15431469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441469 Supplier Thermo Scientific Alfa Aesar Supplier No. H64317.14
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 462-72-6 | C4H8BrF | 155.01 g/mol

1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.

Solubility
Sparingly soluble in water at 25°C 0.48 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-72-6
Molecular Formula C4H8BrF
Molecular Weight (g/mol) 155.01
MDL Number MFCD00209551
InChI Key WMCUHRDQSHQNRW-UHFFFAOYSA-N
PubChem CID 10022
IUPAC Name 1-bromo-4-fluorobutane
SMILES C(CCBr)CF

Specifications

Color Colorless
Density 1.444 g/mL
Boiling Point 134°C to 135°C
Flash Point 45°C (113°F)
Linear Formula FCH2CH2CH2CH2Br
Refractive Index 1.439
Quantity 25 g
UN Number UN1993
Beilstein 1731414
Synonym 4-fluorobutyl bromide, butane, 1-bromo-4-fluoro, butane,1-bromo-4-fluoro, fluorobutyl bromide, acmc-20aoww, 4-bromo-1-fluorobutane, 1-bromo-4-fluoro-butane, 4-01-00-00263 beilstein handbook reference, 1-bromo-4-fluorobutane, 1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane
Solubility Information Sparingly soluble in water at 25°C 0.48g/L.
Formula Weight 155.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-fluorobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.