missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-6-phenylhexane, 97%

Product Code. 11313596
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313596 1 g 1g
11333596 250 mg 250mg
11323596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313596 Supplier Thermo Scientific Alfa Aesar Supplier No. L14568.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 27976-27-8 | C12H17Br | 241.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27976-27-8
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD00236022
InChI Key RAOLIGFNQJMMKW-UHFFFAOYSA-N
Synonym 6-bromohexyl benzene, 1-bromo-6-phenylhexane, benzene, 6-bromohexyl, 1-bromo-6-phenyl-hexane, 6-phenylhexyl bromide, hexane, 1-bromo-6-phenyl, 6-bromanylhexylbenzene, 5-benzylpentyl bromide, 6-bromo-1-phenylhexane
PubChem CID 561788
IUPAC Name 6-bromohexylbenzene
SMILES C1=CC=C(C=C1)CCCCCCBr

Specifications

Boiling Point 115°C to 118°C (6 mmHg)
Refractive Index 1.5295
Quantity 1 g
Beilstein 2207649
Formula Weight 241.18
Percent Purity 97%
Chemical Name or Material 1-Bromo-6-phenylhexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.