missing translation for 'onlineSavingsMsg'
Learn More

1-Bromodecane, 98%

Product Code. 11414190
Change view
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
Product Code. Quantity unitSize
11414190 100 g 100g
11433754 500 g 500g
11443754 2500 g 2500g
11424190 10,000 g 10000g
4 options
This item is not returnable. View return policy

Product Code. 11414190

Brand: Thermo Scientific Alfa Aesar A11340.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-29-8 | C10H21Br | 221.18 g/mol

1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-29-8
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.18
MDL Number MFCD00000221
InChI Key MYMSJFSOOQERIO-UHFFFAOYSA-N
Synonym decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
PubChem CID 8173
IUPAC Name 1-bromodecane
SMILES CCCCCCCCCCBr

Specifications

Melting Point -30°C
Density 1.067
Boiling Point 238°C
Flash Point 94°C (201°F)
Linear Formula CH3(CH2)9Br
Refractive Index 1.456
Quantity 100 g
UN Number UN2810
Beilstein 1735227
Solubility Information Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water.
Formula Weight 221.19
Percent Purity 98%
Chemical Name or Material 1-Bromodecane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromodecane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Bromodecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.