missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromodecane-d21, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromodecane-D21 |
| CAS | 98195-39-2 |
| Molecular Formula | C10 D21 Br |
| Quantity | 0.1 g |
| Synonym | Decane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-d21, 10-bromo- (9 CI), 1-Bromodecane-d21, 1-Bromodecane-D21, 10-Bromodecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-d21, 1-Bromodecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21, 1-Decyl-d21 bromide |
| InChI Formula | InChI=1 S/C10H21Br/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosadeuteriodecane |
| Molecular Weight (g/mol) | 242.3071 |
| Formula Weight | 241.2145 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction