missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane (n-Lauryl Bromide), TRC

Product Code. 30331197
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331197 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331197 Supplier Toronto Research Chemicals Supplier No. TRCB686548100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromododecane (n-Lauryl Bromide)
CAS 143-15-7
Molecular Formula C12H25Br
Quantity 100 g
Synonym 1-Dodecyl Bromide, 12-Bromododecane, Dodecyl Bromide, Lauryl Bromide, NSC 6786, n-Dodecyl Bromide, n-Lauryl Bromide
InChI Formula InChI=1S/C12H25Br/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3
SMILES CCCCCCCCCCCCBr
IUPAC Name 1-bromododecane
Molecular Weight (g/mol) 249.23
Formula Weight 248.114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.