missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%

Product Code. 10625872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10625872 25 g Glass bottle 25g
10753534 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625872 Supplier Thermo Scientific Acros Supplier No. 218390250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 585-71-7 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.3500g/mL
Boiling Point 94.0°C (16.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)Br
Refractive Index 1.5593 to 1.5613
Quantity 25 g
Beilstein 05, 355
Specific Gravity 1.35
Formula Weight 185.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (1-Bromoethyl)benzene
Show More Show Less
compliance-icons
Product Identifier
  • (1-Bromoethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (1-Bromoethyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.