missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromoethyl)benzene, 97%

Product Code. 10753534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753534 100 g Glass bottle 100g
10625872 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753534 Supplier Thermo Scientific Acros Supplier No. 218391000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 585-71-7 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 585-71-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000139
InChI Key CRRUGYDDEMGVDY-UHFFFAOYSA-N
Synonym 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene
PubChem CID 11454
IUPAC Name 1-bromoethylbenzene
SMILES CC(C1=CC=CC=C1)Br

Specifica

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.3500g/mL
Boiling Point 94.0°C (16.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)Br
Refractive Index 1.5593 to 1.5613
Quantity 100 g
Beilstein 05, 355
Specific Gravity 1.35
Formula Weight 185.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (1-Bromoethyl)benzene
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.