missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoheptane, 99%

Product Code. 10656754 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10656754

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-04-9 | C7H15Br | 179.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-04-9
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00000273
InChI Key LSXKDWGTSHCFPP-UHFFFAOYSA-N
Synonym heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane
PubChem CID 12369
IUPAC Name 1-bromoheptane
SMILES CCCCCCCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -58°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 180°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)6Br
Refractive Index 1.4489 to 1.4509
Quantity 1 L
Beilstein 01, 155
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble
Formula Weight 179.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromoheptane, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.